Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Underphophorylated Retinoblastoma-rb-set Mouse anti-Human, FITC, Clone: G99-549, BD

Catalog No. BDB550501 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550501 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550501 Supplier BD Biosciences Supplier No. 550501

Mouse Monoclonal Antibody

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Underphophorylated Retinoblastoma-rb-set
Applications Flow Cytometry
Classification Monoclonal
Clone G99-549
Conjugate FITC
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Includes FITC Mouse Anti-Underphosphorylated Retinoblastoma (Rb) (100 Tests), FITC Mouse IgG1, κ Isotype Control (100 Tests)
Target Species Human
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.