Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rb2 Mouse, Unlabeled, Clone: 10, BD

Catalog No. BDB610261 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610261 50 μg
BDB610262 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610261 Supplier BD Biosciences Supplier No. 610261
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The pRb2 protein shows a high degree of identity with pRb and the related p107 protein. Both pRb and p107 bind to the adenovirus E1A, SV40 large T antigen, and papillomavirus E7 viral proteins. This binding initiates the release of transcription factors which are required for the expression of cell cycle-regulated genes. pRb, pRb2, and p107 interact with a conserved motif in these three viral proteins. In the E1A protein, this area is known as transforming domain 2, which is required for growth activation. The E1A-binding domain in pRb and p107 is a conserved motif known as the “pocket region” which consists of conserved A and B regions separated by non-conserved spacers of different sizes in pRb and p107. It is the pocket regions of pRb and p107 that associate with the E2F transcription factor. pRb2 also contains the conserved pocket region suggesting that it has functional similarities to pRb and p107.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rb2
Applications Western Blot
Classification Monoclonal
Clone 10
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Drosophila, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.