Learn More
Description
The pRb2 protein shows a high degree of identity with pRb and the related p107 protein. Both pRb and p107 bind to the adenovirus E1A, SV40 large T antigen, and papillomavirus E7 viral proteins. This binding initiates the release of transcription factors which are required for the expression of cell cycle-regulated genes. pRb, pRb2, and p107 interact with a conserved motif in these three viral proteins. In the E1A protein, this area is known as transforming domain 2, which is required for growth activation. The E1A-binding domain in pRb and p107 is a conserved motif known as the “pocket region” which consists of conserved A and B regions separated by non-conserved spacers of different sizes in pRb and p107. It is the pocket regions of pRb and p107 that associate with the E2F transcription factor. pRb2 also contains the conserved pocket region suggesting that it has functional similarities to pRb and p107.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | Rb2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 10 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Purification Method | Affinity Purified |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.