Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-RBED1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189848
Description
RBED1 Polyclonal specifically detects RBED1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RBED1 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
C330008I15Rik, ELMO domain-containing protein 3, ELMO/CED-12 domain containing 3, FLJ21977, FLJ35601, liver-specific organic anion transporter 3TM12, LST3, MGC111036, organic anion transporter LST-3b, RBED1, RBM29, RNA binding motif and ELMO domain 1, RNA binding motif and ELMO/CED-12 domain 1, RNA binding motif protein 29, RNA-binding motif and ELMO domain-containing protein 1, RNA-binding motif protein 29, RNA-binding protein 29 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ELMOD3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EWEPRVVSTEVVRAQEEWEAVDTIQPETGSQASSEQPGQLISFSEALQHFQTVDLSPFKKRIQPTIRRTGLAALRHYLFGPPKLHQRLREERDLVLTIAQCGLDSQDPVHG | |
0.1 mL | |
Apoptosis | |
84173 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction