Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514289
Description
This antibody is predicted to react with rat based on sequence homology.
RBM3 is a member of the glycine-rich RNA-binding protein family and a protein with one RNA recognition motif (RRM) domain. Expression of this protein is induced by cold shock and low oxygen tension.
Specifications
| RBM3 | |
| Polyclonal | |
| RBM3 | |
| Putative RNA-binding protein 3; RNA-binding motif protein 3; RNPL; RNPL; RBM3 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 19652, 5935 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| P98179, O89086 | |
| RBM3 | |
| KLH conjugated synthetic peptide between 128-157 amino acids from the C-terminal region of human RBM3 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction