Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA binding motif protein 35A, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89015808 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-015-808 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-015-808 Supplier Abnova Corporation Supplier No. H00054845B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RBM35A protein.

ESPR1 is an epithelial cell-type-specific splicing regulator (Warzecha et al., 2009 [PubMed 19285943]).[supplied by OMIM

Sequence: MTASPDYLVVLFGITAGATGAKLGSDEKELILLFWKVVDLANKKVGQLHEVLVRPDQLELTEDCKEETKIDVESLSSASQLDQALRQFNQSVSNELNIGVGTSFCLCTDGQLHVRQILHPEASKKNVLLPECFYSFFDLRKEFKKCCPGSPDIDKLDVATMTEYLNFEKSSSVSRYGASQVEDMGNIILAMISEPYNHRFSDPERVNYKFESGTCSKMELIDDNTVVRARGLPWQSSDQDIARFFKGLNIAKGGAALCLNAQGRRNGEALVRFVSEEHRDLALQRHKHHMGTRYIEVYKATGEDFLKIAGGTSNEVAQFLSKENQVIVRMRGLPFTATAEEVVAFFGQHCPITGGKEGILFVTYPDGRPTGDAFVLFACEEYAQNALRKHKDLLGKRYIELFRSTAAEVQQVLNRFSSAPLIPLPTPPIIPVLPQQFVPPTNVRDCIRLRGLPYAATIEDILDFLGEFATDIRTHGVHMVLNHQGRPSGDAFIQMKSADRAFMAAQKCHKKNMKDRYVEVFQCSAEEMNFVLMGGTLNRNGLSPPPCKLPCLSPPSYTFPAPAAVIPTEAAIYQPSVILNPRALQPSTAYYPAGTQLFMNYTAYYPSPPGSPNSLGYFPTAANLSGVPPQPGTVVRMQGLAYNTGVKEILNFFQGYQYATEDGLIHTNDQARTLPKEWVCI

Specifications

Antigen RNA binding motif protein 35A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RBM35A protein.
Formulation PBS with no preservative; pH 7.4
Gene RBM35A
Gene Accession No. NM_017697
Gene Alias FLJ20171
Gene Symbols RBM35A
Host Species Mouse
Immunogen RBM35A (NP_060167.2, 1 a.a. ∼ 681 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54845
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.