Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA binding motif, single stranded interacting protein 1, Mouse, Clone: 3F2-2G9, Abnova™

Catalog No. 89000934 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-934 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-934 Supplier Abnova Corporation Supplier No. H00005937M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RBMS1.

This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12. [provided by RefSeq

Sequence: MGKVWKQQMYPQYATYYYPQYLQAKQSLVPAHPMAPPSPSTTSSNNNSSSSSNSGWDQLSKTNLYIRGLPPHTTDQDLVKLCQPYGKIVSTKAILDKTTNKCKGYGFVDFDSPAAAQKAVSALKASGVQAQMAKQQEQDPTNLYISNLPLSMDEQELENMLKPFGQVISTRILRDSSGTSRGVGFARMESTEKCEAVIGHFNGKFIKTPPGVSAPTEPLLCKFADGGQKKRQNPNKYIPNGRPWHREGEAGMTLTYDPTTAAIQNGFYPSPYSIATNRMITQTSITPYIASPVSAYQVQSPSWMQPQPYILQHPGAVLTPSMEHTMSLQPASMISPLAQQMSHLSLGSTGTYMPATSAMQGAYLPQYAHMQTTAVPVEEASGQQQVAVETSNDHSPYTFQPNK

Specifications

Antigen RNA binding motif, single stranded interacting protein 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F2-2G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant RBMS1.
Formulation PBS with no preservative; pH 7.4
Gene RBMS1
Gene Accession No. BC012993
Gene Alias MGC15146/MGC3331/MSSP/MSSP-1/MSSP-2/MSSP-3/SCR2/YC1
Gene Symbols RBMS1
Host Species Mouse
Immunogen RBMS1 (AAH12993, 1 a.a. ∼ 403 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5937
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.