Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBMS1(MSSP-1) Mouse anti-Human, Mouse, Clone: 4D11, MilliporeSigma™

Catalog No. 051360MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-136-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-136-0MI Supplier MilliporeSigma Supplier No. 051360

Mouse Monoclonal Antibody

Anti-RBMS1(MSSP-1) Antibody, clone 4D11 is a Mouse Monoclonal Antibody for detection of RBMS1(MSSP-1) and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMS1(MSSP-1)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4D11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002888.1
Gene Symbols MGC15146, MGC3331, MSSP, MSSP-1, MSSP-2, MSSP-3, MSSP1, SCR2, YC1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresonding to RBMS1 (MSSP-1).
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.