Learn More
RBMY1A1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RBMY1A1 peptide using ARM Technology.
Supplier: Abnova Corporation H00005940K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. Multiple copies of this gene are found in the AZFb azoospermia factor region of chromosome Y and the encoded protein is thought to be involved in spermatogenesis. Most copies of this locus are pseudogenes, although six highly similar copies have full-length ORFs and are considered functional. Four functional copies of this gene are found within inverted repeat IR2; two functional copies of this gene are found in palindrome P3, along with two copies of PTPN13-like, Y-linked. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RBMY1A1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RBMY1A1 | |
RBMY1A1 | |
A synthetic peptide of human RBMY1A1 | |
RUO | |
5940 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
RNA binding motif protein, Y-linked, family 1, member A1 | |
RBMY1A1 | |
MGC181956, RBM1, RBM2, RBMY, YRRM1, YRRM2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.