Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP-jk Rabbit anti-Rat, Mouse, Human, Chicken, Drosophila, Horse, Nematode, Zebrafish, Polyclonal, MilliporeSigma™

Catalog No. AB2284MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2284MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2284MI Supplier MilliporeSigma Supplier No. AB2284

Rabbit Polyclonal Antibody

Anti-RBP-jk Antibody detects level of RBP-jk and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP-Jk
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in PBS containing 0.05% NaN3.
Gene Symbols CBF-1 ; CBF1; IGKJRB; IGKJRB1; KBF2; MGC61669; OTTHUMP00000123440; OTTHUMP00000123441; RBP-J; RBP-JK; RBPJK; RBPSUH; SUH; csl
Host Species Rabbit
Immunogen Synthetic Linear peptide corresponding to the Beta-Trefoil Domain a.a. 194-206 of RBP-jk conjugated to KLH.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005349.2
Target Species Chicken, Drosophila, Equine, Human, Mouse, Nematode, Rat, Zebrafish
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.