Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

retinol binding protein 4, plasma, Mouse, Clone: 4H7, Abnova™

Catalog No. 89000936 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-936 Supplier Abnova Corporation Supplier No. H00005950M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RBP4.

This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells. [provided by RefSeq

Sequence: ERDCRVSSFRVKENFDKARFSGTWYAMAKKDPEGLFLQDNIVAEFSVDETGQMSATAKGRVRLLNNWDVCADMVGTFTDTEDPAKFKMKYWGVASFLQKGNDDHWIVDTDYDTYAVQYSCRLLNLDGTCADSYSFVFSRDPNGLPPEAQKIVRQRQEELCLARQYRLIVHNGYCDGRSERNLL

Specifications

Antigen retinol binding protein 4, plasma
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 4H7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant RBP4.
Formulation PBS with no preservative; pH 7.4
Gene RBP4
Gene Accession No. BC020633
Gene Symbols RBP4
Host Species Mouse
Immunogen RBP4 (AAH20633.1, 19 a.a. ∼ 201 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5950
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.