Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

recombination signal binding protein for immunoglobulin kappa J region, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017765 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-765 Supplier Abnova Corporation Supplier No. H00003516D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human RBPJ protein.

Sequence: MAPVVTGKFGERPPPKRLTREAMRNYLKERGDQTVLILHAKVAQKSYGNEKRFFCPPPCVYLMGSGWKKKKEQMERDGCSEQESQPCAFIGIGNSDQEMQQLNLEGKNYCTAKTLYISDSDKRKHFMLSVKMFYGNSDDIGVFLSKRIKVISKPSKKKQSLKNADLCIASGTKVALFNRLRSQTVSTRYLHVEGGNFHASSQQWGAFFIHLLDDDESEGEEFTVRDGYIHYGQTVKLVCSVTGMALPRLIIRKVDKQTALLDADDPVSQLHKCAFYLKDTERMYLCLSQERIIQFQATPCPKEPNKEMINDGASWTIISTDKAEYTFYEGMGPVLAPVTPVPVVESLQLNGGGDVAMLELTGQNFTPNLRVWFGDVEAETMYRCGESMLCVVPDISAFREGWRWVRQPVQVPVTLVRNDGIIYSTSLTFTYTPEPGPRPHCSAAGAILRANSSQVPPNESNTNSEGSYTNASTNSTSVTSSTATVVS

Specifications

Antigen recombination signal binding protein for immunoglobulin kappa J region
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human RBPJ protein.
Formulation PBS with no preservative; pH 7.4
Gene RBPJ
Gene Accession No. NM_015874.3
Gene Alias CBF1/IGKJRB/IGKJRB1/KBF2/MGC61669/RBP-J/RBPJK/RBPSUH/SUH/csl
Gene Symbols RBPJ
Host Species Rabbit
Immunogen RBPJ (NP_056958.3, 1 a.a. ∼ 487 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 3516
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.