Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RBPMS, Polyclonal

Catalog No. ABN1362MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1362MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1362MI Supplier MilliporeSigma™ Supplier No. ABN1362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects RBPMS Clone: in Human, Mouse, Rat samples, and it is validated for Immunofluorescence, Western Blotting

RNA-binding protein with multiple splicing (UniProt Q93062; also known as Heart and RRM expressed sequence, Hermes, RBP-MS) is encoded by the RBPMS (also known as HERMES) gene (Gene ID 11030) in human. RBPMS is a member of the RRM (RNA recognition motif) family of RNA-binding proteins. Transcriptome-wide study using photoactivatable-ribonucleoside-enhanced crosslinking and immunoprecipitation (PAR-CLIP) in HEK293 cells reveals both exonic mature and intronic pre-mRNA-binding sites, in agreement with the nuclear and cytoplasmic localization of the RRM family proteins. RBPMS is found to be exclusively expressed in retinal ganglion cells (RGCs) and displaced RGCs (dRGCs). RBPMS expression can therefore be utitized as a RGC marker, and the RBPMS gene promoter could be utilized for driving RGC-specific transgene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPMS
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 1% BSA and 0.09 % sodium azide.
Gene Accession No. Q93062
Gene Symbols RBPMS, HERMES
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence from the N-terminal region of human RBPMS.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001008710
Test Specificity Expected to react with all five spliced isoforms (UniProt Q93062-1 through Q93062-5).
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.