Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

recombination signal binding protein for immunoglobulin kappa J region-like, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007189 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-189 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-189 Supplier Abnova Corporation Supplier No. H00011317B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RBPSUHL protein.

In mouse, recombining binding protein L (RBP-L) is a transcription factor that binds to DNA sequences almost identical to that bound by the Notch receptor signalling pathway transcription factor RBP-J. However, unlike RBP-J, RBP-L does not interact with Notch receptors. RBP-L has been shown to activate transcription in concert with Epstein-Barr virus nuclear antigen-2 (EBNA2). The protein encoded by this gene is similar in sequence to the mouse RPB-L protein and Drosophila suppressor of hairless protein. [provided by RefSeq

Sequence: MDPAGAADPSVPPNPLTHLSLQDRSEMQLQSEADRRSLPGTWTRSSPEHTTILRGGVRRCLQQQCEQTVRILHAKVAQKSYGNEKRFFCPPPCVYLSGPGWRVKPGQDQAHQAGETGPTVCGYMGLDSASGSATETQKLNFEQQPDSREFGCAKTLYISDADKRKHFRLVLRLVLRGGRELGTFHSRLIKVISKPSQKKQSLKNTDLCISSGSKVSLFNRLRSQTVSTRYLSVEDGAFVASARQWAAFTLHLADGHSAQGDFPPREGYVRYGSLVQLVCTVTGITLPPMIIRKVAKQCALLDVDEPISQLHKCAFQFPGSPPGGGGTYLCLATEKVVQFQASPCPKEANRALLNDSSCWTIIGTESVEFSFSTSLACTLEPVTPVPLISTLELSGGGDVATLELHGENFHAGLKVWFGDVEAETMYRSPRSLVCVVPDVAAFCSDWRWLRAPITIPMSLVRADGLFYPSAFSFTYTPEYSVRPGHPGVPEPATDADALLESIHQEFTRTNFHLFIQT

Specifications

Antigen recombination signal binding protein for immunoglobulin kappa J region-like
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RBPSUHL protein.
Formulation No additive
Gene RBPJL
Gene Accession No. NM_014276.2
Gene Alias RBP-L/RBPSUHL/SUH/SUHL
Gene Symbols RBPJL
Host Species Mouse
Immunogen RBPSUHL (NP_055091.2, 1 a.a. ∼ 517 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 11317
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.