Learn More
RBX1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RBX1 peptide using ARM Technology.
Supplier: Abnova Corporation H00009978K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes an evolutionarily conserved protein that interacts with cullins. The protein plays a unique role in the ubiquitination reaction by heterodimerizing with cullin-1 to catalyze ubiquitin polymerization. It also may be involved in the regulation of protein turn-over. [provided by RefSeq]
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RBX1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RBX1 | |
RBX1 | |
A synthetic peptide of human RBX1 | |
RUO | |
9978 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ring-box 1 | |
RBX1 | |
BA554C12.1, MGC13357, MGC1481, RNF75, ROC1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.