Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCAN1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5085750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146852SS 0.025 mL
NBP146852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146852SS Supplier Novus Biologicals Supplier No. NBP146852SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

RCAN1 Polyclonal specifically detects RCAN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RCAN1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. P53805, Q9JHG6
Gene Alias ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1
Gene Symbols RCAN1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human RCAN1 (within residues 40-90). [Swiss-Prot# P53805]
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1827
Test Specificity This antibody is specific for isoform 1-1L.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.