Learn More
RCBTB1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RCBTB1 peptide using ARM Technology.
Supplier: Abnova Corporation H00055213K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a protein with an N-terminal RCC1 domain and a C-terminal BTB (broad complex, tramtrack and bric-a-brac) domain. In rat, over-expression of this gene in vascular smooth muscle cells induced cellular hypertrophy. In rat, the C-terminus of RCBTB1 interacts with the angiotensin II receptor-1A. In humans, this gene maps to a region of chromosome 13q that is frequently deleted in B-cell chronic lymphocytic leukemia and other lymphoid malignancies. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RCBTB1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RCBTB1 | |
RCBTB1 | |
A synthetic peptide of human RCBTB1 | |
RUO | |
55213 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1 | |
RCBTB1 | |
CLLD7, CLLL7, GLP, MGC33184, RP11-185C18.1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.