Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCBTB2 (M06A), Mouse anti-Human, Clone: 4G12, Abnova™

Catalog No. 89021068 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-068 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-068 Supplier Abnova Corporation Supplier No. H00001102M06A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RCBTB2.

This gene encodes a member of the RCC1-related GEF family. The N-terminal half of the encoded amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran. [provided by RefSeq

Sequence: TIEPRRLDSLNGKKIACLSYGSGPHIVLATTEGEVFTWGHNAYSQLGNGTTNHGLVPCHISTNLSNKQVIEVACGSYHSLVLTSDGEVFAWGYNNSGQVGSGSTVNQPI

Specifications

Antigen RCBTB2
Applications ELISA
Classification Monoclonal
Clone 4G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RCBTB2.
Formulation ascites with no preservative
Gene RCBTB2
Gene Accession No. NM_001268
Gene Alias CHC1L
Gene Symbols RCBTB2
Host Species Mouse
Immunogen RCBTB2 (NP_001259, 86 a.a. ∼ 194 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1102
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.