Learn More
ring finger and CHY zinc finger domain containing 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RCHY1 protein.
Supplier: Abnova Corporation H00025898D01P
Description
The protein encoded by this gene has ubiquitin-protein ligase activity. This protein binds with p53 and promotes the ubiquitin-mediated proteosomal degradation of p53. This gene is oncogenic because loss of p53 function contributes directly to malignant tumor development. Transcription of this gene is regulated by p53. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MAATAREDGASGQERGQRGCEHYDRGCLLKAPCCDKLYTCRLCHDNNEDHQLDRFKVKEVQCINCEKIQHAQQTCEECSTLFGEYYCDICHLFDKDKKQYHCENCGICRIGPKEDFFHCLKCNLCLAMNLQGRHKCIENVSRQNCPICLEDIHTSRVVAHVLPCGHLLHRGYRCPLCMHSALDMTRYWRQLDDEVAQTPMPSEYQNMTVDILCNDCNGRSTVQFHILGMKCKICESYNTAQAGGRRISLDQQSpecifications
| ring finger and CHY zinc finger domain containing 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human RCHY1 protein. | |
| RCHY1 | |
| ARNIP/CHIMP/DKFZp586C1620/PIRH2/PRO1996/RNF199/ZNF363/hARNIP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001009922.1 | |
| RCHY1 | |
| RCHY1 (NP_001009922.1, 1 a.a. ∼ 252 a.a) full-length human protein. | |
| 100 μg | |
| Ubiquitin/Proteasome | |
| 25898 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.