Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

retinol dehydrogenase 13 (all-trans/9-cis), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-603 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-603 Supplier Abnova Corporation Supplier No. H00112724B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RDH13 protein.

This gene encodes a mitochondrial short-chain dehydrogenase/reductase, which catalyzes the reduction and oxidation of retinoids. The encoded enzyme may function in retinoic acid production and may also protect the mitochondria against oxidative stress. Alternatively spliced transcript variants have been described. [provided by RefSeq

Sequence: MEKCEAAAKDIRGETLNHHVNARHLDLASLKSIREFAAKIIEEEERVDILINNAGVMRCPHWTTEDGFEMQFGVNHLGHFLLTNLLLDKLKASAPSRIINLSSLAHVAGHIDFDDLNWQTRKYNTKAAYCQSKLAIVLFTKELSRRLQGSGVTVNALHPGVARTELGRHTGIHGSTFSSTTLGPIFWLLVKSPELAAQPSTYLAVAEELADVSGKYFDGLKQKAPAPEAEDEEVARRLWAESARLVGLEAPSVREQPLPR

Specifications

Antigen retinol dehydrogenase 13 (all-trans/9-cis)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RDH13 protein.
Formulation PBS with no preservative; pH 7.4
Gene RDH13
Gene Accession No. NM_138412.2
Gene Alias SDR7C3
Gene Symbols RDH13
Host Species Mouse
Immunogen RDH13 (NP_612421.1, 1 a.a. ∼ 260 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 112724
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.