Learn More
RDH13, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RDH13 peptide using ARM Technology.
Supplier: Abnova Corporation H00112724K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a mitochondrial short-chain dehydrogenase/reductase, which catalyzes the reduction and oxidation of retinoids. The encoded enzyme may function in retinoic acid production and may also protect the mitochondria against oxidative stress. Alternatively spliced transcript variants have been described. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| RDH13 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| RDH13 | |
| RDH13 | |
| A synthetic peptide of human RDH13 | |
| RUO | |
| 112724 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| retinol dehydrogenase 13 (all-trans/9-cis) | |
| RDH13 | |
| SDR7C3 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.