Learn More
RDH8, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RDH8 peptide using ARM Technology.
Supplier: Abnova Corporation H00050700K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
All-trans-retinol dehydrogenase (RDH8) is a visual cycle enzyme that reduces all-trans-retinal to all-trans-retinol in the presence of NADPH (Rattner et al., 2000 [PubMed 10753906]). It is a member of the short chain dehydrogenase/reductase family and is located in the outer segments of photoreceptors; hence it is also known as photoreceptor retinol dehydrogenase. It is important in the visual cycle by beginning the rhodopsin regeneration pathway by reducing all-trans-retinal, the product of bleached and hydrolysed rhodopsin (Rando, 2001 [PubMed 11710234]). This is a rate-limiting step in the visual cycle (Saari et al., 1998 [PubMed 9667000]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RDH8 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RDH8 | |
RDH8 | |
A synthetic peptide of human RDH8 | |
RUO | |
50700 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
retinol dehydrogenase 8 (all-trans) | |
RDH8 | |
PRRDH, SDR28C2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.