Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

short chain dehydrogenase/reductase family 16C, member 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018691 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-691 Supplier Abnova Corporation Supplier No. H00195814D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human RDHE2 protein.

RDHE2 belongs to a family of short-chain alcohol dehydrogenases/reductases that catalyze the first and rate-limiting step that generates retinaldehyde from retinol (Matsuzaka et al., 2002 [PubMed 12372410]).[supplied by OMIM

Sequence: MSFNLQSSKKLFIFLGKSLFSLLEAMIFALLPKPRKNVAGEIVLITGAGSGLGRLLALQFARLGSVLVLWDINKEGNEETCKMAREAGATRVHAYTCDCSQKEGVYRVADQVKKEVGDVSILINNAGIVTGKKFLDCPDELMEKSFDVNFKAHLWTYKAFLPAMIANDHGHLVCISSSAGLSGVNGLADYCASKFAAFGFAESVFVETFVQKQKGIKTTIVCPFFIKTGMFEGCTTGCPSLLPILEPKYAVEKIVEAILQEKMYLYMPKLLYFMMFLKSFLPLKTGLLIADYLGILHAMDGFVDQKKKL

Specifications

Antigen short chain dehydrogenase/reductase family 16C, member 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human RDHE2 protein.
Formulation PBS with no preservative; pH 7.4
Gene SDR16C5
Gene Accession No. NM_138969.2
Gene Alias FLJ33105/RDH#2/RDH-E2/RDHE2
Gene Symbols SDR16C5
Host Species Rabbit
Immunogen RDHE2 (NP_620419.2, 1 a.a. ∼ 309 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 195814
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.