Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REA Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07234MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-234-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-234-MI Supplier MilliporeSigma Supplier No. 07234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect REA with Anti-REA Antibody (Rabbit Polyclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REA
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 200 μg of protein A purified rabbit IgG in 200μL of 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
Gene Symbols PHB2; BCAP37; BAP; Prohibitin-2; REA; D-prohibitin; PNAS-141; MGC117268; p22; Bap37
Host Species Rabbit
Immunogen His6-tagged fusion protein corresponding to amino acid residues 88-299 of mouse REA-1/BAP-37/prohibitin-related protein
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_007273
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.