Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RECK, Polyclonal

Catalog No. ST1627100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1627100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1627100UG Supplier MilliporeSigma™ Supplier No. ST1627100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects RECK Clone: in Human samples, and it is validated for Immuno Blotting

The protein encoded by this gene is a cysteine-rich, extracellular protein with protease inhibitor-like domains whose expression is suppressed strongly in many tumors and cells transformed by various kinds of oncogenes. In normal cells, this membrane-anchored glycoprotein may serve as a negative regulator for matrix metalloproteinase-9, a key enzyme involved in tumor invasion and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECK
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Rabbit
Immunogen Full-length, human RECK (aa 1-971)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽125-130kDa RECK protein in HeLa cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.