Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Red Fluorescent Protein Chicken anti-All, Polyclonal, MilliporeSigma™
Chicken Polyclonal Antibody
Supplier: MilliporeSigma AB3528
Description
Anti-Red Fluorescent Protein Antibody is an antibody against Red Fluorescent Protein for use in western blotting.
Specifications
Red Fluorescent Protein | |
Polyclonal | |
Purified immunoglobulin - Ig fraction. Liquid. Buffer = PBS with 0.02% sodium azide | |
Recombinant red fluorescent protein (RFP) highly purified by affinity chromatography on Nickel column. | |
Primary | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Western Blot | |
Unconjugated | |
Chicken | |
100 μg | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction