Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regenerating islet-derived 1 alpha, Mouse, Clone: 4F1, Abnova™

Catalog No. 89000939 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-939 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-939 Supplier Abnova Corporation Supplier No. H00005967M16
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant REG1A.

This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. [provided by RefSeq

Sequence: MNSGNLVSVLTQAEGAFVASLIKESGTDDFNVWIGLHDPKKNRRWHWSSGSLVSYKSWGIGAPSSVNPGYCVSLTSSTGFQKWKDVPCEDKFSFVCKFKN

Specifications

Antigen regenerating islet-derived 1 alpha
Applications ELISA
Classification Monoclonal
Clone 4F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant REG1A.
Formulation PBS with no preservative; pH 7.4
Gene REG1A
Gene Accession No. BC005350
Gene Alias ICRF/MGC12447/P19/PSP/PSPS/PSPS1/PTP/REG
Gene Symbols REG1A
Host Species Mouse
Immunogen REG1A (AAH05350, 67 a.a. ∼ 166 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5967
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.