Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regenerating islet-derived 3 alpha, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017926 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-926 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-926 Supplier Abnova Corporation Supplier No. H00005068D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human REG3A protein.

This gene encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this gene is observed during pancreatic inflammation and liver carcinogenesis. Multiple alternatively spliced transcript variants encoding the same protein have been described for this gene but the full length nature of some transcripts is not yet known. [provided by RefSeq

Sequence: MLPPMALPSVSWMLLSCLMLLSQVQGEEPQRELPSARIRCPKGSKAYGSHCYALFLSPKSWTDADLACQKRPSGNLVSVLSGAEGSFVSSLVKSIGNSYSYVWIGLHDPTQGTEPNGEGWEWSSSDVMNYFAWERNPSTISSPGHCASLSRSTAFLRWKDYNCNVRLPYVCKFTD

Specifications

Antigen regenerating islet-derived 3 alpha
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human REG3A protein.
Formulation PBS with no preservative; pH 7.4
Gene REG3A
Gene Accession No. NM_002580
Gene Alias HIP/INGAP/PAP/PAP-H/PAP1/PBCGF/REG-III/REG3
Gene Symbols REG3A
Host Species Rabbit
Immunogen REG3A (NP_002571.1, 1 a.a. ∼ 175 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5068
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.