Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse RelA/NFkB p65 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems IC5078P
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| RelA/NFkB p65 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65RelA, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5970 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 532301 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q04206 | |
| RELA | |
| E. coli-derived recombinant human RelA/NF kappa B p65 isoform 1 Asn456-Ser551 Accession # Q04206 | |
| 100 Tests | |
| Primary | |
| Detects human and mouse RelA/NF kappa B p65 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction