Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Relaxin 2, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 553850100UG
Description
Specifically detects Relaxin 2 Clone: in Human samples, and it is validated for ELISA, Immunohistochemistry, Radioimmunoassay
Relaxin is a hormone of the insulin-growth factor family that induces ripening of the cervix prior to childbirth. The peptide is composed of two chains, A and B, and is formed from a single-chain prohormone, similar to insulin. Relaxin has also been shown to elicit a powerful vasodilatory response in several target organs. This response is mediated by the stimulation of intrinsic nitric oxide (NO) generation. Additionally, relaxin plays a role in extracellular matrix (ECM) remodeling in many reproductive organs, including the ovary, by upregulating MMP-1, MMP-3, and MMP-9 activity. In the human there are two relaxin genes, H1 and H2, which share 90% sequence homology within their coding region.
Specifications
Relaxin 2 | |
Polyclonal | |
Lyophilized from 50mM Tris, pH 7.5. | |
purified, human relaxin-2 | |
RUO | |
Recognizes the relaxin-2 protein. | |
2°C to 8°C | |
IgG |
ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
Unconjugated | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction