Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RENT1, Polyclonal

Catalog No. DR1015100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DR1015100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DR1015100UG Supplier MilliporeSigma™ Supplier No. DR1015100UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects RENT1 Clone: in Human samples, and it is validated for Immuno Blotting

Eukaryotes have the ability to recognize and degrade transcripts that harbor premature signals for translational termination. Nonsense and frameshift mutations that result in these premature termination codons play a role in many genetic diseases and some cancers. Nonsense-mediated nRNA decay (NMD) is mediated by several factors including rent1/hUpf1, rent2/hUpf2, and hUpf3. When assembled, these proteins form the surveillence complex. Disruption of the mouseRent1 gene has been reported to completely disrupt NMD and result in embryos that re-absorb shortly after implantation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RENT1
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In Tris/citrate phosphate buffer, pH 7.0-8.0.
Host Species Goat
Immunogen a synthetic peptide corresponding to amino acids at the C-terminus of human RENT1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽140kDa RENT1 protein in HeLa cells.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.