Learn More
RALBP1 associated Eps domain containing 2, Mouse, Clone: 4C2, Abnova™
Shop All Abnova Corporation ProductsDescription
The product of this gene is part of a protein complex that regulates the endocytosis of growth factor receptors. The encoded protein directly interacts with a GTPase activating protein that functions downstream of the small G protein Ral. Its expression can negatively affect receptor internalization and inhibit growth factor signaling. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | RALBP1 associated Eps domain containing 2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4C2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant REPS2. |
| Formulation | ascites with no preservative |
| Gene | REPS2 |
| Gene Accession No. | NM_004726 |
| Gene Alias | POB1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.