Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RERE Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526953
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
RERE encodes a member of the atrophin family of arginine-glutamic acid (RE) dipeptide repeat-containing proteins. The encoded protein co-localizes with a transcription factor in the nucleus, and its overexpression triggers apoptosis. A similar protein in mouse associates with histone deacetylase and is thought to function as a transcriptional co-repressor during embryonic development.
Specifications
RERE | |
Polyclonal | |
RERE | |
ARG, ARP, ATN1L, DNB1, arginine-glutamic acid dipeptide repeats protein, atrophin 2, atrophin-1 like protein, atrophin-1 related protein, atrophin-1-like protein, atrophin-1-related protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
473, 68703 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9P2R6, Q80TZ9 | |
RERE | |
KLH conjugated synthetic peptide between 383-409 amino acids from the N-terminal region of human RERE | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction