Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resistin Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB3708MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3708MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3708MI Supplier MilliporeSigma Supplier No. AB3708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Resistin using this Anti-Resistin Antibody validated for use in ELISA, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Resistin
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 20mM PBS containing 0.1% sodium azide.
Gene Symbols RETN; XCP1; MGC126603; RSTN; RETN1; HXCP1; resistin; MGC126609; FIZZ3; ADSF
Host Species Rabbit
Immunogen Synthetic peptide from mouse resistin (aa 83-91).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_020415.2
Target Species Mouse
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.