Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1128MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1128MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1128MI Supplier MilliporeSigma Supplier No. AB1128
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Anti-Respiratory Syncytial Virus Antibody is an antibody against Respiratory Syncytial Virus for use in ELISA, immunofluoresence, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Respiratory Syncytial Virus
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin in PBS (0.01 M, pH 7.2) with 0.1% sodium azide.
Host Species Goat
Immunogen Human RSV isolate
Purification Method Ammonium sulfate precipitation
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.