Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Retinal Pigment Epithelium 65 (RPE65), ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5428/DEL

Description
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
Retinal Pigment Epithelium 65 (RPE65) | |
Unconjugated | |
Bovine RPE microsomal membranes. | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Neuroscience | |
Bovine, Human, Mouse, Xenopus species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction