Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinoblastoma Protein Mouse anti-Human, Unlabeled, Clone: G3-245, BD

Catalog No. BDB554136 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554136 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554136 Supplier BD Biosciences Supplier No. 554136
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the retinoblastoma (Rb) family, including the related proteins p107 and p130, share several properties, including the ability to regulate E2Fdependent transcription and to regulate cell-cycle progression. The Rb gene product is a phosphoprotein that is expressed in most normal cells of vertebrates. Rb acts as a tumor suppressor by providing a cell cycle checkpoint between the G1 and S phases. The active, underphosphorylated form of Rb (Rb or pRb) is primarily found in resting or fully differentiated cells. The activity of Rb is negatively regulated by cyclin dependent kinases, which phosphorylate Rb in late G1. Thus, the hyperphosphorylated form (ppRb) is primarily found in proliferating cells. pRB inactivation is a critical step leading to S-phase commitment at the G1 checkpoint of the cell cycle. In addition, the underphosphorylated form of Rb may bind to viral oncogenes such as SV40 large T Ag, adenoviral EIA and HPV-E7, which may contribute to the transforming activity of these viral oncoproteins. G3-245 was made using a Trp-E-Rb fusion protein as immunogen and recognizes an epitope between amino acids 332-344 (DARLFDHDKTLQ) of the human retinoblastoma protein (pp110-114 Rb). In western blot analysis, Rb migrates as multiple closely-spaced bands between 110-116kda on SDS-PAGE. The bands represent different Rb phosphorylation states where the level of Rb phosphorylation can be cell cycle and/or cell-type dependent (i.e all forms may not be seen in all cell types that express Rb). G3-245 has been reported to recognize human, monkey, mouse, rat, mink and a putative quail Rb. This antibody has also been referred to as Mh-RB-02,20 and mAb-245.

Host Species: Mouse
Clone: G3-245
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Rb aa. 332-344
Formula Weight [Chemical]: 110-116kDa

Bioimaging, Blocking Assay, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Retinoblastoma Protein
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone G3-245
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Rb aa. 332-344
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mink, Monkey, Mouse, Quail, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.