Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Retinoic Acid Receptor, Beta (hRARBeta) Ab-2, Clone: 336, same as H1/H12, Epredia™ LabVision

Catalog No. MS1342PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
200 μL
5 Slides
500 μL
7 mL
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1342PABX 200 μL 1mg/mL
MS1342P1 500 μL 200μg/mL
MS1342P 1 mL 200μg/mL
MS1342P1ABX 100 μL 1mg/mL
MS1342R7 7 mL
MS1342PCS 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1342PABX Supplier Epredia Supplier No. MS1342PABX

Mouse Monoclonal Antibody

Retinoids are metabolites of vitamin A (retinal) and are believed to represent important signaling molecules during vertebrate development and tissue differentiation. Two families of retinoid receptors have been identified. Retinoic acid receptors (RARs) include RARα, RARΒ and RARγ, each of which has a high affinity for all-trans retinoic acids and belongs to the same class of nuclear transcription factors as thyroid hormone receptors, vitamin D3 receptor and ecdysone receptor. The ligand binding domains of the RARs are highly conserved and RAR isoforms are expressed in distinct patterns throught development and in the mature organism. Members of the retinoid X receptor (RXR) family, RXRα, RXRΒ and RXRγ, are activated by 9-cis-retinoic acid, a stereo- and photoisomer of all-trans-RA that is expressed in vivo in both liver and kidney and may represent a widely used hormone.

Host Species: Mouse
Clone: 336; same as H1/H12
Isotype: IgG1/κ
Cross Reactivity: Human. Others-not known.
Epitope: aa 11-25
Immunogen: Synthetic peptide corresponding to aa 11-25 (SPGQILDFYTASPSS) from N-terminus of human RARΒ.
Molecular Weight: 54kDa
Positive Control: HeLa cells. Colon CA.
Cellular Localization: Nuclear

Immunohistochemistry (Formalin/Paraffin)

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Retinoic Acid Receptor Β (hRARΒ) Ab-2
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Synthetic peptide corresponding to aa 11-25 (SPGQILDFYTASPSS) from N-terminus of human RARBeta.
Purification Method HeLa cells, colon CA
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage > 0 degrees
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.