Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Retinoic Acid Receptor Gamma Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN409
Description
Anti-Retinoic Acid Receptor Gamma Antibody is a highly specific rabbit polyclonal antibody, that targets Retinoic Acid Receptor and has been tested in western blotting and immunohistochemistry.
Specifications
Retinoic Acid Receptor Gamma | |
Polyclonal | |
Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunohistochemistry, Western Blot | |
Unconjugated | |
RARG; NR1B3 | |
KLH-conjugated linear peptide corresponding to the C-terminus of human Retinoic Acid Receptor Gamma. | |
100 μg | |
NP_000957 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction