Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Retinoid-Related Orphan Receptor γ T Armenian Hamster anti-Mouse, Clone: RORg2

Catalog No. 04959MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-959-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-959-MI Supplier MilliporeSigma™ Supplier No. 04959
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Detect Retinoid-Related Orphan Receptor γ T using this Anti-Retinoid-Related Orphan Receptor γ T Antibody, clone RORg2 validated for use in western blot, FC and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Retinoid-Related Orphan Receptor γ T
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone RORg2
Conjugate Unconjugated
Formulation Purified hamster monoclonal in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_035411
Gene Symbols RORG, RZRG, NR1F3, RORC, TOR
Host Species Armenian Hamster
Immunogen Histidine-tagged recombinant protein corresponding to mouse Retinoid-Related Orphan Receptor γ T (RORγT).
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.