Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Rex-1 Mouse anti-Human, Mouse, Rat, Clone: 5B4.2, MilliporeSigma™

Catalog No. MAB4316MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4316MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4316MI Supplier MilliporeSigma Supplier No. MAB4316

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Detect Rex-1 with Mouse Anti-Rex-1 Antibody (Mouse Monoclonal Antibody), that has been shown to work in ELISA, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rex-1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 5B4.2
Conjugate Unconjugated
Formulation Purified ascites presented as a liquid in PBS with 0.1% sodium azide.
Gene Accession No. NM_174900
Gene Symbols REX-1, REX1, ZNF754, Zfp-42, ZFP42, hREX-1
Host Species Mouse
Immunogen Recombinant GST-Human fusion protein, human REX-1
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.