Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

replication factor C (activator 1) 3, 38kDaa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018425 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-425 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-425 Supplier Abnova Corporation Supplier No. H00005983D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human RFC3 protein.

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36kDa. This gene encodes the 38kDa subunit. This subunit is essential for the interaction between the 140kDa subunit and the core complex that consists of the 36, 37, and 40kDa subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq

Sequence: MSLWVDKYRPCSLGRLDYHKEQAAQLRNLVQCGDFPHLLVYGPSGAGKKTRIMCILRELYGVGVEKLRIEHQTITTPSKKKIEISTIASNYHLEVNPSDAGNSDRVVIQEMLKTVAQSQQLETNSQRDFKVVLLTEVDKLTKDAQHALRRTMEKYMSTCRLILCCNSTSKVIPPIRSRCLAVRVPAPSIEDICHVLSTVCKKEGLNLPSQLAHRLAEKSCRNLRKALLMCEACRVQQYPFTADQEIPETDWEVYLRETANAIVSQQTPQRLLEVRGRLYELLTHCIPPEIIMKGLLSELLHNCDGQLKGEVAQMAAYYEHRLQLGSKAIYHLEAFVAKFMALYKKFMEDGLEGMMF

Specifications

Antigen replication factor C (activator 1) 3, 38kDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human RFC3 protein.
Formulation PBS with no preservative; pH 7.4
Gene RFC3
Gene Accession No. NM_002915
Gene Alias MGC5276/RFC38
Gene Symbols RFC3
Host Species Rabbit
Immunogen RFC3 (NP_002906.1, 1 a.a. ∼ 356 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5983
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.