Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524620
Description
This antibody is predicted to react with bovine based on sequence homology.
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa.
Specifications
RFC3 | |
Polyclonal | |
RFC3 | |
RFC38, A1 38 kDa subunit, RF-C 38 kDa subunit, RFC, 38 kD subunit, activator 1 38 kDa subunit, activator 1 subunit 3, replication factor C 38 kDa subunit, replication factor C subunit 3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5983, 69263 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
P40938, Q8R323 | |
RFC3 | |
KLH conjugated synthetic peptide between 230-258 amino acids from the C-terminal region of human RFC3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction