Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS1 Polyclonal Antibody, Invitrogen™

RGS1 Antibody, PA1-31563, from Invitrogen™. Species Reactivity: Human, Mouse; Applications: Western Blot.

Supplier:  Thermo Scientific PA131563

Catalog No. PIPA131563


Description

Description

RGS1 Antibody, PA1-31563, from Invitrogen. Species Reactivity: Human, Mouse; Applications: Western Blot.

Regulator of G-protein signalling 1, which negatively regulates G protein-coupled receptor signalling.
Host Species: Goat
Cross Reactivity: Human, Mouse
Isotype: IgG
Immunogen: Peptide with sequence NLLNDLQANSLK, from C Terminus of the protein sequence according to NP_002913
Formulation: 50μg purified antibody (0.5mg/mL) in TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Synonyms: 1R20, BL34, IER1, IR20
Uniprot Accession: Q08116, Q9JL25
Entrez GeneID: 50778, 5996
Gene Symbol: RGS1
PA1-31563 detects RGS1 from human samples. PA1-31563 has been successfully used in Western blot applications. The PA1-31563 immunogen is: Peptide with sequence NLLNDLQANSLK, from C Terminus of the protein sequence according to NP_002913
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RGS1
Polyclonal
Q08116, Q9JL25
RGS1
Peptide with sequence NLLNDLQANSLK, from C Terminus of the protein sequence according to NP_002913
RUO
50778, 5996
Store at -20°C, Avoid Freeze/Thaw Cycles
IgG
Western Blot
Unconjugated
1R20, BL34, IER1, IR20
Goat
50 μg
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.