Learn More
RGS12, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA125694
Description
This gene encodes a member of the 'regulator of G protein signaling' (RGS) gene family. The encoded protein may function as a guanosine triphosphatase (GTPase)-activating protein as well as a transcriptional repressor. This protein may play a role in tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their biological nature has not been determined.
PA1-25694 detects RGS12 from human, mouse, and rat samples.PA1-25694 has been successfully used in Western blot procedures.The PA1-25694 immunogen is a synthetic peptide corresponding to residues 1372-1447 of RGS12.
Specifications
| RGS12 | |
| Polyclonal | |
| Unconjugated | |
| O14924, Q8CGE9, O08774 | |
| RGS12 | |
| Synthetic peptide corresponding to residues 1372-1447 of RGS12. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
| Western Blot | |
| 1mg/mL | |
| RGS12 | |
| RP11-529E10.1, DKFZp761K1617, DKFZp761K1817 | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| 6002 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.