Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regulator of G-protein signaling 5, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016945 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-945 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-945 Supplier Abnova Corporation Supplier No. H00008490B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RGS5 protein.

The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators.[supplied by OMIM

Sequence: MCKGLAALPHSCLERAKEIKIKLGILLQKPDSVGDLVIPYNEKPEKPAKTQKTSLDEALQWRDSLDKLLQNNYGLASFKSFLKSEFSEENLEFWIACEDYKKIKSPAKMAEKAKQIYEEFIQTEAPKEVNIDHFTKDITMKNLVEPSLSSFDMAQKRIHALMEKDSLPRFVRSEFYQELIK

Specifications

Antigen regulator of G-protein signaling 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RGS5 protein.
Formulation PBS with no preservative; pH 7.4
Gene RGS5
Gene Accession No. NM_003617
Gene Alias MST092/MST106/MST129/MSTP032/MSTP092/MSTP106/MSTP129
Gene Symbols RGS5
Host Species Mouse
Immunogen RGS5 (NP_003608.1, 1 a.a. ∼ 181 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8490
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.