Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHEB1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09247MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-247-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-247-MI Supplier MilliporeSigma Supplier No. 09247

Rabbit Polyclonal Antibody

Anti-RHEB1 Antibody detects level of RHEB1 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHEB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100 μg of Protein A purified rabbit polyclonal IgG in 0.02M PBS, 0.25M sodium chloride(pH 7.6) containing 0.1% sodium azide. (Lot 0610041682 provided as ∼13μL at 7.9mg/mL)
Gene Symbols RHEB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide (C-GTAVDVFRRIILEAEK) corresponding to amino acids 155-169 of human Rheb1
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_005605.1
Target Species Human
Content And Storage −20°C for up to 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.