Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Mouse anti-Human, Mouse, Rat, Clone: RET-P1, MilliporeSigma™

Catalog No. MAB5316MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5316MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5316MI Supplier MilliporeSigma Supplier No. MAB5316

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Detect Rhodopsin using this Anti-Rhodopsin Antibody, clone RET-P1 validated for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RET-P1
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 10mM PBS, pH 7.4 with 0.2% BSA and 15mM sodium azide.
Gene Accession No. NM_000539.2
Gene Symbols RHO, rhodopsin, Opsin-2, OPN2, Rhodopsin, RP4, MGC138311, MGC138309
Host Species Mouse
Immunogen Membrane preparation from adult rat retina.
Quantity 500 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.