Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoE/Rnd3 Mouse anti-Human, Mouse, Clone: 4, MilliporeSigma™

Catalog No. 05723MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-723-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-723-MI Supplier MilliporeSigma Supplier No. 05723
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RhoE/Rnd3 Antibody, clone 4 is an antibody against RhoE/Rnd3 for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoE/Rnd3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4
Conjugate Unconjugated
Formulation Mouse monoclonal ascites IgG in buffer containing 0.05% sodium azide before the addition of glycerol to 30%. Liquid at −20deg;C.
Gene Accession No. NM_028810
Gene Symbols RND3, RHOE, RhoE, Rho8, memB, ARHE, RHO8, Rnd3
Host Species Mouse
Immunogen Full length fusion protein corresponding to residues 1-244 of mouse RhoE/Rnd3. Clone 4.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for up to 12 months.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.