Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoG Mouse anti-Human, Mouse, Rat, Clone: 1F3 B3 E5, MilliporeSigma™

Catalog No. 04486MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-486-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-486-MI Supplier MilliporeSigma Supplier No. 04486
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RhoG Antibody, clone 1F3 B3 E5 is an antibody against RhoG for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoG
Applications Western Blot
Classification Monoclonal
Clone 1F3 B3 E5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b in storage buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NM_001665
Gene Symbols RHOG ras homolog gene family, member G (rho G), Rho G, ARHG, MGC125835, MGC125836
Host Species Mouse
Immunogen KLH synthetic peptide encompassing amino acids CQQDGVKEVFAEAVRAVLNPT (RhoG 160-180)
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.